Learn More
Cole-Parmer PVDF Tubing
Chemical- and abrasion-resistant tubing at a low cost
955.00 NOK - 2130.00 NOK
Specifications
Certifications/Compliance | ADCF;Non-Phthalate;RoHS;USP |
---|---|
Overall Length (English) | 50 ft. |
Product Code | Brand | Diameter (English) Inner | Diameter (English) Outer | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) Inner | Diameter (English) Outer | Price | Quantity & Availability | |||||
30053486
|
Cole-Parmer
50119-30 |
5/32 in. | 1/4 in. |
1915.00 NOK
15m |
Estimated Shipment: 14-10-2025
Log in to see stock. |
|||||
30054003
|
Cole-Parmer
50119-27 |
1/8 in. | 3/16 in. |
1420.00 NOK
15m |
Estimated Shipment: 14-10-2025
Log in to see stock. |
|||||
30053793
|
Cole-Parmer
50119-25 |
1/16 in. | 1/8 in. |
955.00 NOK
15m |
Estimated Shipment: 14-10-2025
Log in to see stock. |
|||||
30053029
|
Cole-Parmer
50119-29 |
3/16 in. | 1/4 in. |
1465.00 NOK
15m |
Estimated Shipment: 14-10-2025
Log in to see stock. |
|||||
30053399
|
Cole-Parmer
50119-28 |
1/8 in. | 1/4 in. |
2130.00 NOK
15m |
Estimated Shipment: 14-10-2025
Log in to see stock. |
|||||
Description
Resilient with good fle in., ibility. Does not contain BPA, phthalates, or PVC. Sterilize by autoclave, gamma irradiation, EtO, and chemical sterilants. USP Class VI compliant. Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selectionSpecifications
ADCF;Non-Phthalate;RoHS;USP | |
50 ft. |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Product Title
Cole-Parmer PVDF Tubing