Learn More
Buerkle™ Pump Station for AccuOne/EnergyOne Pump
To aid in transferring small quantities of watery media up to 300 mPas from canisters, barrels and narrow-necked containers from 32mm opening diameter.
2680.00 NOK - 2890.00 NOK
Specifications
No. per Pack | 1 Ea. |
---|---|
Type | Pump Station |
Applications | For dispensing acids and bases |
For Use With | AccuOne/EnergyOne Pump |
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
11991591
|
Buerkle™
5900-3050 |
Immersion depth: 50 cm |
2680.00 NOK
Each |
Estimated Shipment: 22-05-2025
Log in to see stock. |
||||||
11901601
|
Buerkle™
5900-3070 |
Immersion depth: 70 cm |
2785.00 NOK
Each |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
11941581
|
Buerkle™
5900-3100 |
Immersion depth: 100 cm |
2890.00 NOK
Each |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
Description
- Pump station accessory only
- Available for three different submersible lengths
Specifications
1 Ea. | |
Pump Station | |
For dispensing acids and bases | |
AccuOne/EnergyOne Pump |
Product Title
Buerkle™ Pump Station for AccuOne/EnergyOne Pump