missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTHLH Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1457.58 NOK - 3943.39 NOK
Specifications
Antigen | PTHLH |
---|---|
Concentration | 0.27 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16857453
|
Proteintech
10817-1-AP-20UL |
20 μL |
1457.58 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16847453
|
Proteintech
10817-1-AP-150UL |
150 μL |
3943.39 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a member of the parathyroid hormone family. This hormone regulates endochondral bone development and epithelial-mesenchymal interactions during the formation of the mammary glands and teeth. This hormone is involved in lactation possibly by regulating the mobilization and transfer of calcium to the milk. The receptor of this hormone, PTHR1, is responsible for most cases of humoral hypercalcemia of maligcy. Four alternatively spliced transcript variants encoding two distinct isoforms have been observed. There is also evidence for alternative translation initiation from non-AUG (CUG and GUG) start sites, in-frame and downstream of the initiator AUG codon, to give rise to nuclear forms of this hormone.Specifications
PTHLH | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
P12272 | |
5744 | |
PTHLH Fusion Protein Ag1133 | |
Primary | |
-20°C | |
PTHLH |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
HHM, PLP, PTH rP, PTHLH, PTHR, PTHRP, PTHrP[107 139] | |
PTHLH | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title