Learn More
Fisherbrand™ PTFE Tweezers, Fine Square
Fisherbrand™ PTFE Tweezers, Fine Square are inert and non-contaminating
Brand:  Fisherbrand™ 027.0200
 View more versions of this product
Product Code. 16296021
  • 158.00 NOK / Each
Only null left
Estimated Shipment: 11-08-2025
Add to basket
Description
Description
  • Total Chemical Resistance
  • High Purity
  • Temperature Range -200°C to +280°C
  • Non contaminating
  • Easy to clean
Specifications
Specifications
PTFE
200 mm
Fisherbrand™ PTFE Tweezers, Fine Square
1/Ea.
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ PTFE Tweezers, Fine Square > Length: 200mm