Learn More
Fisherbrand™ PTFE Tweezers, Fine Square
Fisherbrand™ PTFE Tweezers, Fine Square are inert and non-contaminating
Brand:  Fisherbrand™ 027.0100
 View more versions of this product
Product Code. 16256021
  • 457.00 NOK / Pack of 5
Only null left
Estimated Shipment: 07-08-2025
Add to basket
Description
Description
  • Total Chemical Resistance
  • High Purity
  • Temperature Range -200°C to +280°C
  • Non contaminating
  • Easy to clean
Specifications
Specifications
PTFE
100 mm
Fisherbrand™ PTFE Tweezers, Fine Square
1/Ea.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ PTFE Tweezers, Fine Square > Length: 100mm