Learn More
Fisherbrand™ PTFE Tweezers, Fine Sharp
Fisherbrand™ PTFE Tweezers, Fine Sharp are inert and non-contaminating
Brand: Fisherbrand™ 027.0101
Description
- Total Chemical Resistance
- High Purity
- Temperature Range -200°C to +280°C
- Non contaminating
- Easy to clean
Specifications
PTFE | |
100 mm | |
Fisherbrand™ PTFE Tweezers, Fine Sharp | |
1/Ea. |
Product Content Correction
Product Title
Fisherbrand™ PTFE Tweezers, Fine Sharp > Length: 100mm