Learn More
Cowie Technology™ PTFE Straight Through Stopcocks
Cowie Technology™ PTFE Straight Through Stopcocks feature a pure PTFE body and plug and combine all the desirable properties of the Ultimate Laboratory Stopcock.
Brand: Cowie Technology™ 016.002
This item is not returnable.
View return policy
Description
- Medium vacuum pressure performance
- Unbreakable
- Chemically inert
- Grease free
- Supplied with borosilicate glass sidearms
Specifications
Straight Through | |
Straight Through Stopcock | |
Autoclavable | |
1 Each | |
PTFE |
2 mm | |
PTFE | |
8 mm | |
Borosilicate glass sidearms |
Product Content Correction
Product Title
Cowie Technology™ PTFE Straight Through Stopcocks > Bore Size: 2 mm