Learn More
Wheaton™ PTFE/Silicone Septa
Brand:  Wheaton™ W240584A
 View more versions of this product
Product Code. 10210042
  • 3470.00 NOK / Pack of 100
Only null left
Estimated Shipment: 18-07-2025
Specifications
15 mm
Autoclavable
Septa
PTFE, Silicone
100 Pk.
SDS
Documents Certificates
Promotions
Promotions
Product Content Correction
Product Title
Wheaton™ PTFE/Silicone Septa > Outer Diameter: 15mm