Learn More
Bochem™ Sharp Tip Forceps with Guide Pin
With a guide pin to help keep the tips aligned.
Brand: Bochem™ 1811
This item is not returnable.
View return policy
Description
- Manufactued from PTFE-coated stainless steel
- Sharp tips for excellent precision
Specifications
Sharp | |
Stainless Steel | |
Forceps | |
Guide pin |
1 Pack | |
130 mm | |
PTFE coating |
Product Content Correction
Product Title
Bochem™ Sharp Tip Forceps with Guide Pin > Length: 130mm