Learn More
Glindemann™ PTFE Sealing Rings for taper joints (laboratory glassware)
For grease-free sealing of ground joints and plugs, extra tight and extra clean. The alternative to ground collar, grip collar sleeve and stopper made of PTFE.
Brand:  Glindemann™ K45
Encompass_Preferred
 View more versions of this product
Product Code. 10686901
  • 686.77 NOK / Pack of 50
Only null left
Estimated Shipment: 21-07-2025
Description
  • Air leak rate <10-8 to 10-6 mbar l/s (high vacuum, overpressure).
  • Solvent leak rate in containers <0.1 to 0.03mg/d.
  • Temperature resistant from -200 to +300°C.
  • The elastic ring (0.07mm thick) adheres to the ground joint without a groove, is reusable and fits to ground joint clamps.
  • Other sizes on request.
Specifications
Specifications
PTFE
NS 45
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Glindemann™ PTFE Sealing Rings for taper joints (laboratory glassware)