Learn More
- Home
- Products
- Stoppers and Closures
- Bottle Caps
- 11786898
Favorite item to keep tabs on it!
Saved!
Go to My Lists
You have reached the limit of 350 favorited items.
Manage Your Favorites
This item is not returnable.
View return policy
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Product Code. 11786898
Brand: Duran™ 1165525
Estimated Shipment: 27-03-2025
Log in to see stock.
Log in to see stock.
Add to basket
Add to basket
This item is not returnable.
View return policy
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Description
- GL 45
- PTFE seal
Specifications
Specifications
Material | PTFE |
Product Type | Screw Cap |
Closure Liner | PTFE |
Closure Size | GL 45 |
Product Suggestions
You may be interested in these alternatives
-
-
Product Code. 10222161
-
Product Code. 10650901
Customers who viewed this item also viewed This information does not imply a recommendation or representation of any kind and any action taken upon the information provided is strictly at your own risk. The products shown have not been qualified by Fisher Scientific for any particular purpose, application or use and it is the responsibility of the purchaser to ensure that the performance of the product is suitable for the purchasers specific use or intended application.
Viewing 1-5 of 9
-
Catalog No. 12347178
kr 2 293,20 / Pack of 10 -
Catalog No. 15486113
kr 1 190,00 / Pack of 10 -
Catalog No. 17916792
kr 3 505,00 / Pack of 10 -
Catalog No. 17383863
kr 283,59 / Pack of 100 -
Catalog No. 10571013
kr 196,52 / Pack of 100 -
Catalog No. 11580535
kr 309,00 / Pack of 100 -
Catalog No. 10222161
kr 1 279,20 / Pack of 10 -
Catalog No. 11762332
kr 317,20 / Pack of 10 -
Catalog No. 10166642
kr 831,00 / Pack of 2
Product Content Correction
Product Title
Duran™ PTFE Screw Cap >