Learn More
Fisherbrand™ PTFE Plain Stirrer Shaft
Fisherbrand™ PTFE Plain Stirrer Shafts have a PTFE encapulated stainless steel core with exposed end. They are a fully molded construction featuring a PTFE roto molded to a fully PTFE encapsulated steel reinforced drive shaft.
367.00 NOK - 1920.00 NOK
Product Code | Brand | Dimensions (L x W) | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Diameter (Metric) | Price | Quantity & Availability | |||||
16265891
|
Fisherbrand™
005.100830EX |
300 x 8 mm | 5 mm |
374.00 NOK
Each |
Estimated Shipment: 25-08-2025
Log in to see stock. |
|||||
16275891
|
Fisherbrand™
005.100850EX |
500 x 8 mm | 5 mm |
443.00 NOK
Each |
Estimated Shipment: 19-09-2025
Log in to see stock. |
|||||
16285891
|
Fisherbrand™
005.101030EX |
300 x 10 mm | 6.35 mm |
367.00 NOK
Each |
Estimated Shipment: 19-09-2025
Log in to see stock. |
|||||
16295891
|
Fisherbrand™
005.101050EX |
500 x 10 mm | 6.35 mm |
442.00 NOK
Each |
Estimated Shipment: 25-08-2025
Log in to see stock. |
|||||
16205901
|
Fisherbrand™
005.101250EX |
500 x 12 mm | 6.35 mm |
780.00 NOK
Each |
Estimated Shipment: 19-09-2025
Log in to see stock. |
|||||
16215901
|
Fisherbrand™
005.101265EX |
650 x 12 mm | 6.35 mm |
835.00 NOK
Each |
Estimated Shipment: 19-09-2025
Log in to see stock. |
|||||
16225901
|
Fisherbrand™
005.101275EX |
750 x 12 mm | 6.35 mm |
840.00 NOK
Each |
Estimated Shipment: 19-09-2025
Log in to see stock. |
|||||
16235901
|
Fisherbrand™
005.1016100EX |
1000 x 16 mm | 10 mm |
1340.00 NOK
Each |
Estimated Shipment: 19-09-2025
Log in to see stock. |
|||||
16245901
|
Fisherbrand™
005.1016750EX |
750 x 16 mm | 10 mm |
1250.00 NOK
Each |
Estimated Shipment: 19-09-2025
Log in to see stock. |
|||||
16255901
|
Fisherbrand™
005.1019100EX |
1000 x 19 mm | 10 mm |
1920.00 NOK
Each |
Estimated Shipment: 19-09-2025
Log in to see stock. |
|||||
16265901
|
Fisherbrand™
005.1019750EX |
750 x 19 mm | 10 mm |
1650.00 NOK
Each |
Estimated Shipment: 19-09-2025
Log in to see stock. |
|||||
Description
- PTFE with Steel Core
- 50mm Exposed End
- Chemically Inert
- Easy to clean
- No cross contamination
- Non scratch
- Use up to 280° C
Specifications
PTFE Plain Stirrer Shaft | |
1 |
Product Title
Fisherbrand™ PTFE Plain Stirrer Shaft