Learn More
BRAND™ PTFE Magnetic Stirring Bars Retriever
Manufactured from PTFE. BRAND™ PTFE Magnetic Stirring Bars Retriever has a magnetic core that is fully sealed to prevent leeching.
Brand:  BRAND™ 137700
236.97 NOK valid until 2025-07-13
Use promo code "24135" to get your promotional price.
 View more versions of this product
Product Code. 10018310
  • 268.00 NOK / Each
Elevate Your Beverage Testing with Precision Solutions
Use promo code 24135 at checkout
View Available Promotions
Only null left
Estimated Shipment: 10-07-2025
Description
  • Manufactured from PTFE.
  • Magnetic core is fully sealed to prevent leeching.
  • Manufactured from PTFE.
  • Magnetic core is fully sealed to prevent leeching.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
8 mm
150 mm
PTFE
1
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BRAND™ PTFE Magnetic Stirring Bars Retriever > Length (Metric): 150 mm