Learn More
Fisherbrand™ PTFE Magnetic Stir Bars with Pivot Ring
Used in containers with bases that are slightly curved or uneven.
Brand:  Fisherbrand™ 001.350
Product Code. 16255811
  • 427.00 NOK / Pack of 10
Only null left
Estimated Shipment: 01-08-2025
Add to basket
Description
Description
  • Used in containers with bases that are slightly curved or uneven
Specifications
Specifications
Autoclavable
8 mm
1/Pk.
Containers & Vessels / Stirrer Hot Plates
50 mm
Cylinder
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ PTFE Magnetic Stir Bars with Pivot Ring > Diameter: 8mm; Length: 50mm