missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Industrial Flat Tip pH Electrodes, High Temperature, PTFE

Ideal for continuous in-line monitoring and for solutions containing aggressive chemicals. Hanna Instruments™ Industrial Flat Tip pH Electrodes, High Temperature, PTFE virtually eliminates deposits that can foul the electrode, significantly reducing necessary maintenance.

Brand:  Hanna Instruments™ HI1006-3005

Product Code. 12981830

  • 11570.00 NOK / Each
Estimated Shipment: 15-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Hanna presents combination pH electrodes incorporating over 20 years of electrode manufacturing experience.

  • Self-cleaning flat tip sensor for significantly reduced maintenance
  • Internal matching pin avoids typical problems caused by grounding loop current
  • 3/4 in. NPT external thread on both ends for easy installation
  • Annular non-clogging PTFE junction for testing solutions with high content of suspended solids or for high pressure installation
  • Electrodes are wired with the standard BNC connector
  • Specialized glass can withstand high temperature process and streams
  • Non amplified electrode for short distance measurements supplied with 5m (16.4 ft.) cable
Specifications

Specifications

BNC
High temperature processes and streams
16.4 ft. Cable
High temperature glass, PTFE junction
Industrial Flat Tip pH Electrode
Cable
5 m Cable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hanna Instruments™ Industrial Flat Tip pH Electrodes, High Temperature, PTFE > Connector Type: BNC

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.