Learn More
Bohlender™ PTFE Cylindrical Magnetic Stirring Bar
Brand: Bohlender™ C350-08
This item is not returnable.
View return policy
Specifications
FDA Certified | |
Alnico 5 magnetic core | |
10 mm | |
1/Pk. |
Cylindrical magnetic stirring bars offer excellent centering and smooth running characteristics | |
3 mm | |
PTFE | |
Cylinder |
Product Content Correction
Product Title
Bohlender™ PTFE Cylindrical Magnetic Stirring Bar > Length: 10mm