Learn More
Poulten & Graf GmbH PTFE Adaptor for FORTUNA™ Dispenser with Thread
Complete a FORTUNA™ Dispenser with an adapter that comes in a wide selection of threads. Poulten & Graf GmbH PTFE Adaptor for FORTUNA™ Dispenser with Thread enables smaller containers to be supported on top the existing reservior.
Brand:  Poulten & Graf GmbH 101 09155
 View more versions of this product
Product Code. 10585352
  • 1870.00 NOK / Each
Only null left
Estimated Shipment: 15-07-2025
Add to basket
Description
Description
  • Suitable for OPTIFIX and POLYFIX dispenser
  • Made of PTFE (Teflon™)
  • Adapter with threads
  • Can be used for acids and alkaline solutions that do not attack polypropylene
Specifications
Specifications
Adapters for OPTIFIX and POLYFIX Dispensers
White
PTFE
1
Autoclavable
Adapters for OPTIFIX and POLYFIX dispensers
OPTIFIX and POLYFIX dispensers
GL 55
Product Suggestions
Product Suggestions
Videos
Videos
  • Poulten Graf FORTUNA OPTIMAT 3
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Poulten Graf FORTUNA OPTIMAT 3
    SDS
    Documents
    Documents
    Certificates
    Promotions
    Promotions
    Product Content Correction
    Product Title
    Poulten & Graf GmbH PTFE Adaptor for FORTUNA™ Dispenser with Thread > ThreadSize: GL 55