Learn More
Poulten & Graf GmbH PTFE Adaptor for FORTUNA™ Dispenser with Thread
Complete a FORTUNA™ Dispenser with an adapter that comes in a wide selection of threads. Poulten & Graf GmbH PTFE Adaptor for FORTUNA™ Dispenser with Thread enables smaller containers to be supported on top the existing reservior.
Brand: Poulten & Graf GmbH 101 09160
This item is not returnable.
View return policy
Description
- Suitable for OPTIFIX and POLYFIX dispenser
- Made of PTFE (Teflon™)
- Adapter with threads
- Can be used for acids and alkaline solutions that do not attack polypropylene
Specifications
Adapters for OPTIFIX and POLYFIX Dispensers | |
White | |
PTFE | |
1 |
Autoclavable | |
Adapters for OPTIFIX and POLYFIX dispensers | |
OPTIFIX and POLYFIX dispensers | |
GL 60 |
Videos
-
Poulten Graf FORTUNA OPTIMAT 3
Poulten and Graf Parts for Dispensers
File size: 622.9 kB
Product Content Correction
Product Title
Poulten & Graf GmbH PTFE Adaptor for FORTUNA™ Dispenser with Thread > ThreadSize: GL 60