Learn More
Poulten & Graf GmbH PTFE Adaptor for FORTUNA™ Dispenser with Thread
Complete a FORTUNA™ Dispenser with an adapter that comes in a wide selection of threads. Poulten & Graf GmbH PTFE Adaptor for FORTUNA™ Dispenser with Thread enables smaller containers to be supported on top the existing reservior.
Brand:  Poulten & Graf GmbH 101 09133
 View more versions of this product
Product Code. 10301562
  • 682.00 NOK / Each
Description
  • Suitable for OPTIFIX and POLYFIX dispenser
  • Made of PTFE (Teflon™)
  • Adapter with threads
  • Can be used for acids and alkaline solutions that do not attack polypropylene
Specifications
Specifications
Adapters for OPTIFIX and POLYFIX Dispensers
White
PTFE
1
Autoclavable
Adapters for OPTIFIX and POLYFIX dispensers
OPTIFIX and POLYFIX dispensers
GL 33
Product Suggestions
Product Suggestions
Videos
Videos
  • Poulten Graf FORTUNA OPTIMAT 3
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Poulten & Graf GmbH PTFE Adaptor for FORTUNA™ Dispenser with Thread > ThreadSize: GL 33