Learn More
Poulten & Graf GmbH PTFE Adaptor for FORTUNA™ Dispenser with Thread
Complete a FORTUNA™ Dispenser with an adapter that comes in a wide selection of threads. Poulten & Graf GmbH PTFE Adaptor for FORTUNA™ Dispenser with Thread enables smaller containers to be supported on top the existing reservior.
Brand:  Poulten & Graf GmbH 101 09152
 View more versions of this product
Product Code. 10289331
  • 1870.00 NOK / Each
Description
  • Suitable for OPTIFIX and POLYFIX dispenser
  • Made of PTFE (Teflon™)
  • Adapter with threads
  • Can be used for acids and alkaline solutions that do not attack polypropylene
Specifications
Specifications
Adapters for OPTIFIX and POLYFIX Dispensers
White
PTFE
1
Autoclavable
Adapters for OPTIFIX and POLYFIX dispensers
OPTIFIX and POLYFIX dispensers
GL 52
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Poulten & Graf GmbH PTFE Adaptor for FORTUNA™ Dispenser with Thread > ThreadSize: GL 52