Learn More
Thermo Scientific™ PT100 External Probe
For Use With: Cimarec and Supernuova Hotplates and Hotplate Stirrers
Brand: Thermo Scientific™ CIC0001447
674.78 NOK valid until 2025-08-24
Use promo code "24140" to get your promotional price.
Description
For Use With: Cimarec and Supernuova Hotplates and Hotplate Stirrers
- Composed of AISI 316Ti, a titanium-stabilized version of the 316 stainless steel
- AISI 316Ti is a type of austenitic chromium-nickel stainless steel specifically designed to resist intergranular corrosion at high temperatures
Specifications
PT100 External probe, 0-400°C, Stainless steel, for all sizes of Cimarec+ and SuperNuova+ Hotplates and Hotplate Stirrers | |
0°C to 400°C | |
Digital |
Cimarec and Supernuova Hotplates and Hotplate Stirrers | |
Probe | |
Stainless Steel |
Promotions
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59%