Learn More
Heidolph™ Pt 1000 temperature sensor for Magnetic Stirrer
For use with MR Hei-End, MR Hei-tec, MR Hei-Connect and MR Hei-Standard magnetic stirrers
Brand: Heidolph™ 509-67910-00
1337.89 NOK valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Description
- For use with MR Hei-End, MR Hei-tec, MR Hei-Connect and MR Hei-Standard magnetic stirrers
Specifications
For use with MR Hei-End, MR Hei-tec, MR Hei-Connect and MR Hei-Standard magnetic stirrer |
Promotions
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
Days
58Hours
11Minutes
55Seconds
0 -
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24156Expires: 2025-06-30
Days
3Hours
11Minutes
55Seconds
0