Learn More
PSMG2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10959-1-AP-20UL
 View more versions of this product
Product Code. 16847693
  • 1696.32 NOK / 20µL
Explore more special offers
Description
Description
Chaperone protein which promotes assembly of the 20S proteasome as part of a heterodimer with PSMG1. The PSMG1-PSMG2 heterodimer binds to the PSMA5 and PSMA7 proteasome subunits, promotes assembly of the proteasome alpha subunits into the heteroheptameric alpha ring and prevents alpha ring dimerization.
Specifications
Specifications
PSMG2
Polyclonal
Unconjugated
PSMG2
CLAST3, HCCA3, HsT1707, MDS003, PAC 2, PAC2, PSMG2, TNFSF5IP1
Rabbit
Antigen Affinity Chromatography
RUO
107047, 291539, 56984
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q969U7, Q9EST4
Psmg2
PSMG2 Fusion Protein Ag1400
20 μL
Primary
Mouse, Rat, Human