missing translation for 'onlineSavingsMsg'
Learn More

PSMD4 Mouse anti-Human, Clone: 3G1A1, Proteintech

Mouse Monoclonal Antibody

1611.84 NOK - 4593.76 NOK

Specifications

Antigen PSMD4
Clone 3G1A1
Concentration 1 mg/mL
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16887815
View Documents Promotion Details
Proteintech
66179-1-IG-20UL
20 μL
1611.84 NOK
20µL
Please to purchase this item. Need a web account? Register with us today!
16877815
View Documents Promotion Details
Proteintech
66179-1-IG-150UL
150 μL
4593.76 NOK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody is specific to PSMD4.

This gene encodes a subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This subunit functions as guanine nucleotide exchange factor. It is reported that this subunit interacts with HIV-1 Tat, and thus it represses the translation of host-cell, but not HIV-1, mRNAs. Several alternatively spliced transcript variants encoding multiple isoforms have been found for this gene.
Specifications

Specifications

PSMD4
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
AF, AF 1, Antisecretory factor 1, ASF, MCB1, PSMD4, pUB R5, Rpn10, S5A
PSMD4
IgG1
Protein G
Antibody
3G1A1
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
Mouse
Human
P55036
5710
PSMD4 Fusion Protein Ag7010
Primary
-20°C
PSMD4
Videos
SDS
Documents

Documents

Product Certifications