missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSAPL1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55128-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody is specific to PSAPL1.
May activate the lysosomal degradation of sphingolipids.Specifications
PSAPL1 | |
Polyclonal | |
Unconjugated | |
PSAPL1 | |
prosaposin like 1, PSAPL1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
768239 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.49 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6NUJ1 | |
PSAPL1 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |