Learn More
Cytiva Whatman™ Protein Prep for ӒKTA™ Chromatography Systems
The Protein Prep syringe filter containing the regenerated cellulose membrane is the optimal choice for filtration prior to chromatography with ӒKTA™ systems.
Brand:  Cytiva 10463103
 View more versions of this product
Product Code. 15861838
  • 4375.00 NOK / Pack of 150
Only null left
Estimated Shipment: 04-07-2025
Description
  • Suitable for numerous applications in pharmaceutical, environmental, biotechnology, food/beverage and agricultural testing laboratories
  • Heat sealed without the use of glues or sealants
  • Designed for optimal preparation of protein samples prior to chromatography
  • High yield protein purification and accurate protein analysis both depend on a quality sample being introduced to the chromatography column. The Protein Prep syringe filter has been designed specifically to optimize these outcomes
  • Filtration of samples prior to runs demonstrated to reduce column degrading particulate matter
  • Hydrophilic membrane made of regenerated cellulose exhibits low protein binding for sample preservation
  • Lot-to-lot certified to have low levels of extractables that could otherwise interfere with work
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    0.2 μm
    150 Pieces
    13 mm
    Documents
    One moment while we fetch your results.
    Certificates
    Promotions
    Promotions
    Product Content Correction
    Product Title
    Cytiva Whatman™ Protein Prep for ӒKTA™ Chromatography Systems > Diameter Metric: 13 mm, PoreSize: 0.2 μm