Learn More
Applied Biosystems™ Protein Expression Sample Prep Kit
Shop All Applied Biosystems Products
Click to view available options
Quantity:
1000 reactions
Unit Size:
Each
Description
The Applied Biosystems™ Protein Expression Sample Preparation Kit contains the reagents required for the preparation of target and control cell lysates for use with TaqMan™ Protein Expression Assay experiments. The Cell Lysis Reagent is a buffered detergent solution, which is combined with user-supplied protease and phosphatase inhibitors, and used to lyse adherent or nonadherent cultured cells under nondenaturing conditions. The Cell Resuspension Buffer is used to resuspend the cells before adding the lysis reagent and to dilute the lysates in preparation for the assay-binding step.
Specifications
Specifications
Type | Cell Lysis Buffer |
Quantity | 1000 reactions |
Content And Storage | Contains Cell Lysis Reagent (2x) (3 x 1 mL tubes) and Cell Resuspension Buffer (1 x 25 mL bottle). Store at 2 to 8°C. |
Product Line | TaqMan™ |
Documents
Product Information
Manuals
Quick Reference Card: TaqMan Protein Assays Sample Prep
Version: 4449771 Rev B
File size: 656.7 kB
Getting Started Guide - ProteinAssist Software - Data analysis software for TaqMan Protein Assays
Version: 4457491 Rev B
File size: 6.7 MB
Quick Reference Card: TaqMan® Protein Assays (English )
Version: 16 Feb 2011
File size: 661.6 kB
Manual: Real-Time PCR Systems TaqMan® Protein Assays Chemistry Guide (English )
Version: 13 Apr 2011
File size: 3.3 MB
Protocol: TaqMan® Protein Assays Sample Prep and Assay (English )
Version: 16 Feb 2011
File size: 2.6 MB
Protocol: Protein Expression Sample Preparation Kit
Version: A
File size: 49.9 kB
Manual: ProteinAssist™ Software Getting Started Guide (English )
Version: 08 Nov 2010
File size: 6.7 MB
Specification Sheets
Brochures & Specifications: TaqMan® Protein Assays Brochure
File size: 2.1 MB
Brochures & Specifications: TaqMan Protein Assays Product Overview
File size: 346.7 kB
Brochures & Specifications: TaqMan Protein Expression Assays
File size: 957.6 kB
Scientific Resources
Posters
Novel Protein Expression Assays using qPCR for the Detection and Relative Quantification of Protein Markers in Stem Cells
File size: 230.4 kB
Conference Poster: Novel Protein Expression Assays using qPCR for the Detection and Relative Quantitation of Protein Markers in Stem Cell
File size: 230.4 kB
An integrated qPCR approach for monitoring gene and protein expression in human pluripotent & differentiated cells
File size: 170.9 kB
Detection of clinically relevant proteins in human testicular germ cell tumors
File size: 243.8 kB
Application Notes
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
2828751 | Certificate of Analysis | 29/01/2025 | 4405443 |
2838193 | Certificate of Analysis | 29/01/2025 | 4405443 |
2782168 | Certificate of Analysis | 10/08/2023 | 4405443 |
2747923 | Certificate of Analysis | 21/06/2023 | 4405443 |
2571716 | Certificate of Analysis | 21/10/2022 | 4405443 |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Product Title
Applied Biosystems™ Protein Expression Sample Prep Kit >