missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Protein A, Recombinant

Recombinant Protein A is useful for the study of cell surface antigens and receptors, and for detecting antibody-secreting cells.

Brand:  MP Biomedicals™ 08987051

Product Code. 11410175

  • 6435.00 NOK / 5mg
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Recombinant protein A binds immunoglobulins (primarily IgG) at the Fc region with high affinity.
  • In some cases, IgM and IgA will also bind. This property provides a rapid, simple and economical method for purification and analysis of a broad-spectrum of antibodies.
  • Fractionation of IgG subclasses can also be performed using recombinant protein A.
  • Because of its high affinity for immunoglobulins of many species and its low level of non-specific binding, recombinant protein A is useful for the study of cell surface antigens and receptors, and for detecting antibody-secreting cells.
  • Recombinant protein A is produced in an E. coli strain which contains the gene for mature protein A from Staphylococcus aureus Cowan I strain.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Protein A Sepharose
Immunoassays
0°C to 5°C
5 mg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.