missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Protective Coverall 4565
Protective Coverall 4565 has desirable comfort features and provides a strong barrier to certain chemicals, liquid sprays and particles.
4530.00 NOK
Specifications
Product Type | Cleanroom Coveralls |
---|---|
Gender | Unisex |
Material | Polypropylene |
Color | White |
Disposable | Single-use |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
17855676
|
3M™
7100256145 |
Small |
4530.00 NOK
Pack of 20 |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||||
17845666
|
3M™
7100256062 |
Medium |
4530.00 NOK
Pack of 20 |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||||
17805666
|
3M™
7100256144 |
Large |
4530.00 NOK
Pack of 20 |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||||
17865666
|
3M™
7100256143 |
X-Large |
4530.00 NOK
Pack of 20 |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||||
17875666
|
3M™
7100256140 |
2X-Large |
4530.00 NOK
Pack of 20 |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||||
17845676
|
3M™
7100256139 |
3X-Large |
4530.00 NOK
Pack of 20 |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||||
17875676
|
3M™
7100256138 |
4X-Large |
4530.00 NOK
Pack of 20 |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||||
Description
- Wear 3M™ Protective Coverall 4565 for an ideal barrier to hazardous particles, limited liquid splashes and sprays to Type 4
- Fresh white coverall 4565 has recognizable red taped seams and meets a variety of standards, including chemical protective clothing EN 14126 Infective agents, EN 1073-2 Nuclear particulate resistance, EN 1149, EN ISO 13688, Type 6 Liquid Splash EN 13034 and Type 5 Dust EN ISO 13982-1, and Type 4 Liquid Spray EN 14605
- Taped seams reduce skin exposure points through tearing and reduced risk of inward leakage by chemicals or particles
- Offers Til class 2 in Particulate Radioactive Contamination EN1073-2, meaning it has very good particle hold out as well as liquid spray resistance
- Made with a non-microporous PE laminate for chemical protection with permeation-tested resistance against certain chemicals
- Knitted cuffs, elasticated waist and ankles make the wearer more comfortable and increase freedom of movement, while the ultra-low linting material reduces environmental contamination
- Anti-static treatment meeting EN 1149, which reduces static discharge and risk of spark and ignition
Specifications
Cleanroom Coveralls | |
Polypropylene | |
Single-use | |
Elastic | |
Hooded | |
2-way Zipper | |
Bloodborne Pathogens | |
Controlled Environments |
Unisex | |
White | |
Elastic | |
Knit | |
3-panel | |
Non-sterile | |
None | |
Class 10-100,000 (ISO 4-8) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3M™ Protective Coverall 4565