Learn More
Honeywell™ PROTECT (i)XTREM S3 Shoes
Honeywell™ PROTECT (i)XTREM S3 Shoes are constructed of water-resistant full grain leather.
1290.00 NOK
Specifications
Certifications/Compliance | EN ISO 20345:2011 S3 HRO HI CI SRC |
---|---|
Color | Black |
Material | Leather |
Sole Material | Polyurethane and nitrile |
Toe Style | Metal-free |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
15816585
|
Honeywell™
6246204-37/7 |
37 |
1290.00 NOK
1 pair |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
15826585
|
Honeywell™
6246204-38/7 |
38 |
1290.00 NOK
1 pair |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
15836585
|
Honeywell™
6246204-39/7 |
39 |
1290.00 NOK
1 pair |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
15846585
|
Honeywell™
6246204-40/7 |
40 |
1290.00 NOK
1 pair |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
15856585
|
Honeywell™
6246204-41/7 |
41 |
1290.00 NOK
1 pair |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
15866585
|
Honeywell™
6246204-42/7 |
42 |
1290.00 NOK
1 pair |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
15876585
|
Honeywell™
6246204-43/7 |
43 |
1290.00 NOK
1 pair |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
15886585
|
Honeywell™
6246204-44/7 |
44 |
1290.00 NOK
1 pair |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
15896585
|
Honeywell™
6246204-45/7 |
45 |
1290.00 NOK
1 pair |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
15806595
|
Honeywell™
6246204-46/7 |
46 |
1290.00 NOK
1 pair |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
15816595
|
Honeywell™
6246204-47/7 |
47 |
1290.00 NOK
1 pair |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
15826595
|
Honeywell™
6246204-48/7 |
48 |
1290.00 NOK
1 pair |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
Description
- Water resistant grain full leather
- Ergonomic collar and padded tongue for an optimized comfort
- Gusset to avoid water and dust penetration
- Breathable 3D lining
- Derby style for a better fitting in all sizes
- Toe cap metal free
- Metal free antiperforation insert
- Injected PU / Nitrile sole
- (i)NTELLIGENT wear control
- EN ISO 20345:2011 S3 HRO HI CI SRC
Specifications
EN ISO 20345:2011 S3 HRO HI CI SRC | |
Leather | |
Metal-free |
Black | |
Polyurethane and nitrile |
Product Title
Honeywell™ PROTECT (i)XTREM S3 Shoes