missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ ProteCol™ PEEK Unions

Precision engineered for capillary LC. Trajan Scientific and Medical ProteCol™ PEEK Unions offer complete versatility for connecting capillary tubing.

Brand:  Trajan Scientific and Medical™ 222011

 View more versions of this product

Product Code. 15209223

  • 1525.00 NOK / Pack of 2
Only null left
Estimated Shipment: 24-06-2025
Add to basket
This item is not returnable. View return policy

Description

Description

  • Leak free connections
  • Replacement parts such as nuts and ferrules
Specifications

Specifications

Union
Size: 0.36 mm to 0.36 mm
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Trajan Scientific and Medical™ ProteCol™ PEEK Unions > Size: 0.36mm to 0.36mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.