Learn More
Trajan Scientific and Medical™ ProteCol™ PEEK Unions
Precision engineered for capillary LC. Trajan Scientific and Medical ProteCol™ PEEK Unions offer complete versatility for connecting capillary tubing.
Brand:  Trajan Scientific and Medical™ 212010
 View more versions of this product
Product Code. 13595410
  • 1525.00 NOK / Each
Only null left
Estimated Shipment: 04-07-2025
Add to basket
Description
Description
  • Leak free connections
  • Replacement parts such as nuts and ferrules
Specifications
Specifications
Union
Size: 1.59 mm to 0.79 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Trajan Scientific and Medical™ ProteCol™ PEEK Unions > Size: 1/16 in. to 1/32 in.