missing translation for 'onlineSavingsMsg'
Learn More

Proteasome 20S alpha 6 Rabbit anti-Human, Mouse, Rat, Clone: S01-4I2, Novus Biologicals™

Rabbit Monoclonal Antibody

2690.00 NOK - 5435.00 NOK

Specifications

Antigen Proteasome 20S alpha 6
Clone S01-4I2
Dilution Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18351377
View Documents
Bio-Techne
NBP3-15085-25UL
25 μg
2690.00 NOK
25µL
Please to purchase this item. Need a web account? Register with us today!
18325322
View Documents
Bio-Techne
NBP3-15085-100UL
100 μg
5435.00 NOK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Proteasome 20S alpha 6 Monoclonal antibody specifically detects Proteasome 20S alpha 6 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications

Specifications

Proteasome 20S alpha 6
Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence
Monoclonal
Purified
RUO
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
5687
IgG
Affinity-purified
S01-4I2
Western Blot, ELISA, Immunofluorescence
Unconjugated
Rabbit
Human, Mouse, Rat
EC 3.4.25.1, IOTA, Macropain iota chain, macropain subunit iota, MGC22756, MGC2333, MGC23846, Multicatalytic endopeptidase complex iota chain, P27K, p27KPROS-27, PROS2727 kDa prosomal protein, prosomal P27K protein, proteasome (prosome, macropain) subunit, alpha type, 6, Proteasome iota chain, proteasome subunit alpha type-6, proteasome subunit iota
A synthetic peptide of human Proteasome 20S alpha 6 (Uniprot#: P60900)
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.