missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propoconazole, Tilt, SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
Propiconazole (Tilt) | 60207-90-1 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
CAS | 60207-90-1, 67-64-1 |
Packaging | 2mL amber prescored glass ampule |
CAS Min % | 0.1, 99.9 |
Quantity | 1 mL |
CAS Max % | 0.1, 99.9 |
Concentration or Composition Notes | Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
Molecular Formula | C15H17Cl2N3O2 |
InChI Key | STJLVHWMYQXCPB-UHFFFAOYSA-N |
SMILES | CCCC1COC(O1)(CN2C=NC=N2)C3=C(C=C(C=C3)Cl)Cl |
For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
Show More |
Safety and Handling


Product Identifier
- Propiconazole (Tilt)
- Danger
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : acetone, propiconazole
Product intended for laboratory use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Propoconazole, Tilt, SPEX CertiPrep™ >
Spot an opportunity for improvement?Share a Content Correction