Learn More
Propazine-D7, Honeywell Fluka™
Click to view available options
Quantity:
10 mg
Description
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
- Free expert technical support
Specifications
Specifications
CAS | 1655498-05-7 |
Packaging | Glass Bottle |
Quantity | 10 mg |
Molecular Formula | C9D7H9ClN5 |
Molecular Weight (g/mol) | 236.75 |
Grade | Analytical Standard |
For Use With (Application) | Herbicides |
Physical Form | Crystalline |