Learn More
Heidolph™ Instruments Hei-VAP Automatic Evaporation plus Package, EU-plug
The Hei-VAP Automatic Evaporation plus Package features a motor lift, Woulff bottle, condenser, vacuum pump, and tubing set.
Brand:  Heidolph™ Instruments 574-38310-00
111031.79 NOK valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Additional Details : Weight : 20.00000kg
Product Code. 17249039
  • 131680.00 NOK / Each
Estimated Shipment: 16-08-2024
to see stock.
Add to basket
Description
Description
  • Hei-VAP Expert Control with motor lift and coated vertical glassware set G3B
  • Vacuum pump Rotavac Vario Control
  • Condenser for Rotavac Vario
  • Tubing set
  • Woulff bottle
  • AUTOaccurate-sensor
Specifications
Specifications
Motor Lift Rotary Evaporator
EU-plug