Learn More
Contec ProChlor SATWIPES System
Point-of-use SATWIPE system, sterile ProChlor is a revolutionary sporicide achieving a 100% kill against spores in under 1 min.
Brand: Contec XSWPP02PC
7068.90 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Add 700ml of ProChlor to the dry roll, 5 mins before use, and use the presaturated wipes within 8 hours.
- Meets requirements of Aseptic Transfer Guidelines
- Log 5 kill against Bacillus subtilis in 60 seconds
- Fast acting so suitable for transfer disinfection
- Fast and easy to use
- Very low residue, saving time on residue removal
- Double bagged in linear tear packaging
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
230602453 | Certificate of Quality | 22/06/2024 | XSWPP02PC |
220402034X | Certificate of Quality | 02/06/2023 | XSWPP02PC |
220201988X | Certificate of Quality | 19/04/2023 | XSWPP02PC |