Learn More
Invitrogen™ eBioscience™ ProcartaPlex Human TIMP-1 Simplex
Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.
Brand:  Invitrogen™ EPX01A-12018-901
Product Code. 15552197
  • 2770.00 NOK / 96 tests
Explore more special offers
Includes:
  • concentrated Magnetic Beads coated with monoclonal antibody to human TIMP-1
  • human TIMP-1 Standard (lyophilized)
  • Biotin-Conjugate (50x) anti-human TIMP-1 monoclonal antibody
  • Universal Assay Buffer 10x
Description
Description
  • Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
  • ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
  • It is recommended to dilute TIMP-1 samples when measured in serum or plasma
  • The recommended sample dilution factor is 1:20
TRUSTED_SUSTAINABILITY
Specifications
Specifications
96 Tests
Multiplex Immunoassay
Serum, Plasma: 25 μL; CCS: 50 μL
Human
Cell Culture Supernatant, Plasma, Serum
Tissue inhibitor of metalloproteinases, EPA