missing translation for 'onlineSavingsMsg'
Learn More

PRKACG Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1696.32 NOK - 4796.50 NOK

Specifications

Antigen PRKACG
Concentration 0.35 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16856825
View Documents
Proteintech
55346-1-AP-20UL
20 μL
1696.32 NOK
20µL
Please to purchase this item. Need a web account? Register with us today!
16846825
View Documents
Proteintech
55346-1-AP-150UL
150 μL
4796.50 NOK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cyclic AMP-dependent protein kinase (PKA) consists of two catalytic subunits and a regulatory subunit dimer. This gene encodes the gamma form of its catalytic subunit. The gene is intronless and is thought to be a retrotransposon derived from the gene for the alpha form of the PKA catalytic subunit.
Specifications

Specifications

PRKACG
Western Blot
Unconjugated
Rabbit
Mouse, Human
P22612
5568
Peptide
Primary
-20°C
PRKACG
0.35 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
KAPG, PKA C gamma, PKACg, PRKACG
PRKACG
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.