Learn More
Invitrogen™ PRIMER URACIL 200 NMOL
Custom Primer
Brand:  Invitrogen™ 10623031
104.25 NOK valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Product Code. 10032652
  • 139.00 NOK / Each
Explore more special offers
Description
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Uracil Primer, 200 nmol
Uracil Primer
200 nmol