Learn More
Invitrogen™ PRIMER 5' FAM 10 UMOL
Custom Primer
Brand:  Invitrogen™ 10627057
7228.00 NOK valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Product Code. 10779554
Description
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5' FAM Primer, 10μmol
5' FAM Primer
5' Labeled
10 μmol