Learn More
Tocris Bioscience™ Pregnenolone sulfate sodium salt
NMDA potentiator; also GABAA inhibitor and TRPM3 activator
Brand: Tocris Bioscience™ 5376/50
Description
Pregnenolone sulfate sodium salt is a highly potent neurosteroid with multiple actions. Enhances intracellular Ca2+ upon NMDA receptor-mediated synaptic activity (EC50 ∽ 2 pM) and potentiates NMDA responses. Inhibitor at GABAA receptors (IC50 values are 0.4 to >300 μM, depending on specific subunit conformation). Activates CREB phosphorylation, augments LTP and cognitive behavior in rat memory models. Also activates TRPM3 channels.Specifications
50 mg |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only