missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prealbumin/transthyretin Mouse anti-Human, Mouse, Clone: 1H8C4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66108-1-IG-20UL
This item is not returnable.
View return policy
Description
Transthyretin, one of the three prealbumins including alpha-1-antitrypsin, transthyretin and orosomucoid. Transthyretin is a carrier protein; it transports thyroid hormones in the plasma and cerebrospinal fluid, and also transports retinol (vitamin A) in the plasma. The protein consists of a tetramer of identical subunits. More than 80 different mutations in this gene have been reported; most mutations are related to amyloid deposition, affecting predomitly peripheral nerve and/or the heart, and a small portion of the gene mutations is non-amyloidogenic. The diseases caused by mutations include amyloidotic polyneuropathy, euthyroid hyperthyroxinaemia, amyloidotic vitreous opacities, cardiomyopathy, oculoleptomeningeal amyloidosis, meningocerebrovascular amyloidosis, carpal tunnel syndrome, etc.Specifications
Prealbumin/transthyretin | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P02766, P07309 | |
TTR | |
Prealbumin/transthyretin Fusion Protein Ag19232 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG2a |
Western Blot, Immunohistochemistry (Paraffin) | |
1H8C4 | |
Unconjugated | |
TTR | |
ATTR, HsT2651, PALB, Prealbumin, Prealbumin/transthyretin, TBPA, transthyretin, TTR | |
Mouse | |
Protein A | |
RUO | |
22139, 7276 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction