missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP4C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | PPP4C |
---|---|
Concentration | 0.27 mg/mL |
Applications | Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16856533
|
Proteintech
10262-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16846533
|
Proteintech
10262-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is one subunit of a trimeric complex, forming a highly conserved transcription factor that binds to CCAAT motifs in the promoter regions in a variety of genes. Subunit A associates with a tight dimer composed of the B and C subunits, resulting in a trimer that binds to DNA with high specificity and affinity. The sequence specific interactions of the complex are made by the A subunit, suggesting a role as the regulatory subunit. In addition, there is evidence of post-transcriptional regulation in this gene product, either by protein degradation or control of translation. Further regulation is represented by alternative splicing in the glutamine-rich activation domain, with clear tissue-specific preferences for the two isoforms.Specifications
PPP4C | |
Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
P60510, P97470, Q5BJ92 | |
171366, 5531, 56420 | |
PPP4C Fusion Protein Ag0356 | |
Primary | |
-20°C | |
PPP4C |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
PP X, PP4, PP4C, PPH3, PPP4, PPP4C, PPX, Protein phosphatase X | |
Ppp4c | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |