missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPIL3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15671-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the cyclophilin family. Cyclophilins catalyze the cis-trans isomerization of peptidylprolyl imide bonds in oligopeptides. They have been proposed to act either as catalysts or as molecular chaperones in protein-folding events. Alternative splicing results in multiple transcript variants.Specifications
PPIL3 | |
Polyclonal | |
Unconjugated | |
PPIL3 | |
Cyclophilin J, Cyclophilin like protein PPIL3, CYPJ, PPIase, PPIL3, Rotamase PPIL3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
301432, 53938, 70225 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q812D3, Q9D6L8, Q9H2H8 | |
PPIL3 | |
PPIL3 Fusion Protein Ag8137 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction