Learn More
VACUUBRAND™ Power Supply
Brand:  VACUUBRAND™ 20612409
 View more versions of this product
Product Code. 11960552
  • 1805.00 NOK / Each
Only null left
Estimated Shipment: 12-08-2025
Specifications
Power Supply
24 VDC
Certificates
Promotions
Promotions
Product Content Correction
Product Title
VACUUBRAND™ Power Supply > Voltage: 24VDC