Learn More
Invitrogen™ Power Blotter XL Welcome Pack
Get rapid and efficient semi-dry protein transfer with the Invitrogen™ Power Blotter.
Brand:  Invitrogen™ PB0113
22344.00 NOK valid until 2025-06-30
Use promo code "24108" to get your promotional price.
Product Code. 15853984
  • 29400.00 NOK / 1 set
Nurture Your Progress
Use promo code 24108 at checkout
View Available Promotions
Only null left
Estimated Shipment: 04-07-2025
Add to basket
Includes: Power Blotter Station; Power Blotter Cassette XL; Power Blotter Select Transfer Stacks, Nitrocellulose, Regular; Power Blotter Pre-cut Membranes and Filters, Nitrocellulose, Regular; Power Blotter 1-Step Transfer Buffer
Description
Description
The Invitrogen™ Power Blotter System is designed specifically for rapid semi-dry transfer of proteins from polyacrylamide gels to nitrocellulose or PVDF membranes in 5 to 12 minutes. The Power Blotter features an integrated power supply optimized to enable consistent and high-efficiency protein transfer.
The Power Blotter achieves highly efficient transfer in 7 minutes when used with Power Blotter Select Transfer Stacks or 10 minutes with the Power Blotter 1-Step Transfer Buffer. The instrument is also effective for standard 30- to 60-minute semi-dry transfer protocols based on traditional buffers.
Features:
  • Efficient— high transfer efficiency with a broad range of protein sizes compared to conventional semi-dry or wet (tank) transfer methods
  • Fast—5 to 12 minute transfer when used with Power Blotter Select Transfer Stacks or Power Blotter 1-Step Transfer Buffer
  • Integrated power supply— seamless operation between control unit and cassette; no need for external power supply
  • Easy-touch programming— convenient pre-programmed transfer methods or create, save, and run customized transfer methods
  • Flexible— allows for simultaneous transfer
  • Versatile—compatible with Power Blotter Select Transfer Stacks, Pierce 1-Step Transfer Buffer for rapid blotting programs or Towbin transfer buffer for conventional semi-dry transfer methods
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Novex Bolt gels, NuPage Protein gels
Nitrocellulose
Room Temperature
Power Blotter
1 kit
• Power Blotter Station
• Power Blotter Cassette XL
• Power Blotter Select Transfer Stacks, nitrocellulose, regular-size
• Power Blotter Pre-cut Membranes and Filters, nitrocellulose, regular-size
• Power Blotter 1-Step Transfer Buffer
Semi-Dry
SDS
Documents Certificates
Promotions
Promotions
  • Invitrogen , ABI Applied Biosystems , Thermo Scientific , Affymetrix eBioscience
    Exclusive savings to help your research thrive.
    Promo Code: 24108
    Expires2025-06-27
Product Content Correction
Product Title
Invitrogen™ Power Blotter XL Welcome Pack > Includes: Power Blotter Cassette XL; Size: Cassette size - XL