missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POU5F2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26362-1-AP-150UL
This item is not returnable.
View return policy
Description
Transcription factor that binds preferentially to the octamer motif (5′-ATGTTAAT-3′). May exert a regulatory function in meiotic events that are required for terminal differentiation of male germ cell.Specifications
POU5F2 | |
Polyclonal | |
Unconjugated | |
POU5F2 | |
POU5F2, SPRM 1, SPRM1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
134187 | |
-20°C | |
Liquid |
Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8N7G0 | |
POU5F2 | |
POU5F2 Fusion Protein Ag23803 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction