Learn More
Memmert™ Potential-Free Contact (24V/2A) W/Socket To Namur Ne 28 For Combined Error Message, For Un30Pa
Brand:  Memmert™ UN30paH6
Product Code. 16314417
  • 3445.00 NOK / Each
Estimated Shipment: 01-08-2025
Explore more special offers
Add to basket
Includes: Socket 3-Pin
Specifications
Models VO, VOcool, HCP, TTC, CTC, U, UF TS, UNpa, S, I, ICP, IPP, IPS, HPP, ICH, 108
Potential-free Contact
24 V
2 A
Product Title
Memmert™ Potential-Free Contact (24V/2A) W/Socket To Namur Ne 28 For Combined Error Message, For Un30Pa >