Learn More
Memmert™ Potential-Free Contact (24 V / 2 A) with Socket to NAMUR NE 28 for Combined Error Message for UN30
Potential-free contact for combination error message (e.g. supply failure, sensor fault, fuse).
Brand: Memmert™ UN30H6
Description
- Accessory
- For combination error message
Specifications
UN30 Oven | |
Socket to NAMUR NE 28 | |
2 A |
Potential-free Contact | |
24 V |